Learn More
Hirschmann™ Amber Glass Stoppers
Product Code. 30090417
Click to view available options
Standard Taper Joint:
NS 10/19
NS 12/21
NS 14/23
NS 19/26
NS 24/29
NS 29/32
NS 34/35
NS 45/40
NS 7/16
Unit Size:
Each
Pack of 10
Product Code. 30090417
Brand: Hirschmann™ 8902375
Only null left
Add to basket
Add to basket
Light weight amber colored hollow glass stopper for the protection of UV-sensitive media.
  • Acc. to DIN 12 252
Specifications
For Use With (Equipment) For tight sealing of standard ground vessels
Quantity 10/Pk.
Standard Taper Joint NS 19/26
Material Glass
Product Type Hexagonal Head Stopper