Learn More
Hirschmann™ Pipetting Pliers
Click to view available options
:
Pliers
Description
- Especially suitable for the so-called Delbruck method, because contamination caused by direct finger contact with the outside of the capillary is avoided
- Spreading to the reagent with falsification of analysis is reliably prevented
- Provided in packs of 10
Specifications
Specifications
| Type | Pliers/Tongs |
| For Use With (Application) | Secure holding of micropipettes |
| Quantity | 10/Pk. |