missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ BTB1

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
BTB1 is a selective and ATP-competitive mitotic kinesin Kif18A inhibitor (IC50 = 1.7 μM); reversibly inhibits microtubule-stimulated Kif18A ATPase activity. Displays selectivity against a panel of other mitotic kinesins.
Specifications
Specifications
| Quantity | 50 mg |
| Product Type | BTB1 |
| Purity | >99% |
| Molecular Weight (g/mol) | 297.71 |
| Inhibitors | BTB1 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction