missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ BTD S-sulfenic acid probe

Product Code. 30042613
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
This item is not returnable. View return policy

Product Code. 30042613

Brand: Tocris Bioscience™ 8059/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemoproteomic reagent for targeting S-sulfenic acids on SulfenQ platform, alkyne handle for click chemistry

BTD S-sulfenic acid probe is a nucleophilic fragment that can selectively modify sulfenic acid residues in proteins, and can be used as a tool to identify ligandable S-sulfenation sites. Covalently modifies oxidized cysteines on proteins. Quantitatively labels hyper-reactive sulfenic acids at low BTD concentrations, less reactive sites exhibit concentration-dependent increases in probe labeling. Incorporates an alkyne handle for click chemistry and chemoproteomic profiling.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-(4-Pentyn-1-yl)-1H-2,1-benzothiazin-4(3H)-one 2,2-dioxide
CAS 2095485-22-4
Quantity 10 mg
Target General Laboratory Research Reagent
Molecular Formula C13H13NO3S
Purity 0.98
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.