Learn More
BTX™ Microslides for Electrofusion Chambers
Click to view available options
Size (English) Gap:
0.019 in.
0.039 in.
0.12 in.
0.39 in.
Electrode Shape:
Rectangular Electrodes
Round Wire Electrodes
Field Type:
Divergent
Homogeneous
Volume (Metric):
20 μL
40 μL
650 μL
2 mL
Unit Size:
Each
Pack of 10
Description
- Allow alignment and fusion to be observed under a microscope
- Microslide 450 has two stainless steel tubes mounted on a glass slide and generates a divergent field
- Microslide 453 has two stainless steel bars that can be set apart at 3.2 or 10 mm gaps to provide homogenous fields
Specifications
Specifications
| For Use With (Application) | Embryo Manipulation, Nuclear Transfer, Cell Fusion, Hybridoma Production, Plant Protoplast Fusion |
| For Use With (Equipment) | ECM 830, Gemini X2, ECM 2001 Generator |
| Material | Glass chamber, Stainless Steel electrode |
| Display | Color |
| Electrode Shape | Round Wire Electrodes |
| Field Type | Divergent |
| Includes | Glass Microslides with round wire electrodes, 1.0 mm gap, 40 μl, Pkg/10 |
| Quantity | 10 Pk. |
| Size (English) Gap | 0.039 in. |
| Size (Metric) Gap | 1 mm |
| Show More |