Learn More
BUCHI Beaker and Drying Flasks for Rotary Evaporators
Shop All Buchi Products
Click to view available options
Volume (Metric):
500 mL
1000 mL
1500 mL
2000 mL
Type:
Beaker Drying Flask
Beaker Flask
Powder Drying Flask
Unit Size:
Each
Description
- For use with the rotary evaporators
- Available in multiple sizes with different standard joints
Specifications
Specifications
| Material | Borosilicate Glass (3.3) |
| Type | Beaker Drying Flask |
| Temperature (Metric) Operating | 60°C |
| Volume (Metric) | 1500 mL |