Learn More
BUCHI Glass Columns C-690
Brand:  BUCHI 044046
Additional Details : Weight : 3.60000kg
 View more versions of this product
Product Code. 12749485
  • 2215.00€ / Each
Estimated Shipment: 26-04-2024
to see stock.
Specifications
49 mm
920 mm
Borosilicate Glass
Preparative Separations Together with The C-600 Sepacore™ Line of Chromatography Separators
290 psi
Column