Learn More
BUCHI Glass Columns C-690
Brand:  BUCHI 044050
Additional Details : Weight : 5.38000kg
 View more versions of this product
Product Code. 12779485
  • 3795.00€ / Each
Estimated Shipment: 08-04-2024
to see stock.
Specifications
70 mm
920 mm
Borosilicate Glass
Preparative Separations Together with The C-600 Sepacore™ Line of Chromatography Separators
218 psi
Column