Learn More
BUCHI Glass Columns C-690
Product Code. p-4701648 Shop All Buchi Products
Click to view available options
Length (Metric):
460 mm
Diameter (Metric):
26 mm
Max. Pressure:
580 psi
Unit Size:
Each
Product Code. 11967208
Brand: BUCHI 044037
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Specifications
Diameter (Metric) 26 mm
For Use With Preparative Separations Together with The C-600 Sepacore™ Line of Chromatography Separators
Length (Metric) 460 mm
Max. Pressure 580 psi
Material Borosilicate Glass
Type Glass Column