Learn More
BUCHI Heating Bath Top Cover
Shop All Buchi Products
Click to view available options
For Use With (Equipment):
Heating bath B-491, B-100
Unit Size:
Each
Specifications
Specifications
| Description | Top Cover |
| For Use With (Equipment) | Heating bath B-491, B-100 |
| For Use With (Application) | Save energy and to reduce evaporation of heating media |