Learn More
BUCHI Large Capacity Evaporating Flasks
Standard glass flasks for use with various Rotavapor™ Evaporator models
Brand:  BUCHI 027010
Additional Details : Weight : 3.82000kg
 View more versions of this product
Product Code. 12733546
  • 2565.00€ / Each
Estimated Shipment: 30-06-2021
Add to basket
Description
Description
  • Inert industrial glass (borosilicate 3.3, ISO/DIN 3585)
  • Tension relieved at 540°C
Specifications
Specifications
R-152 rotary evaporator
10 L
Standard Glass
Evaporating Flask