missing translation for 'onlineSavingsMsg'
Learn More

BUCHI Calibration and Verification Standards for BUCHI Melting Point Instruments

Product Code. 10532511 Shop All Buchi Products
Click to view available options
:
Calibration Kit; Model: B-540/545
This item is not returnable. View return policy

Product Code. 10532511

Brand: BUCHI 037454

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Includes

Vanillin, Caffeine, Phenacetin

Utilize the embedded firmware and software on your BUCHI melting point apparatus to calibrate and verify your instrument at any time. Comprehensive IQ/OQ procedures including certified calibration and verification compounds for GLP/GMP laboratories.

Utilize the embedded firmware and software on your BUCHI Melting Point apparatus to calibrate and verify your instrument at any time.

  • Comprehensive IQ/OQ procedures including certified calibration and verification compounds for GLP/GMP laboratories
  • Guided calibrations and verifications provide easy and reliable results
  • Ensures compliance from 10°C to 400°C

Specifications

Product Type Calibration Kit
For Use With (Equipment) BUCHI B-540/545 Melting Point apparatus
Includes Vanillin, caffeine, phenacetin
compliance-icons compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Calibration Kit for Melting Point
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 2
  • Acute toxicity Category 4
  • Oxidising solid Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Serious eye damage/eye irritation Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H301 + H311 + H331-Toxic if swallowed, in contact with skin or if inhaled.
  • H315-Causes skin irritation.
  • H302-Harmful if swallowed.
  • H272-May intensify fire; oxidiser.
  • H373-May cause damage to organs.
  • H319-Causes serious eye irritation.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P264-Wash thoroughly after handling.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P321-Specific treatment (see on this label).
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P270-Do not eat, drink or smoke when using this product.
  • P220-Keep/Store away from clothing/combustible materials.
  • P330-Rinse mouth.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P361 + P364-Take off immediately all contaminated clothing and wash it before reuse.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
  • P405-Store locked up.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P370+P378-In case of fire: Use CO2, extinguishing powder or water spray. to extinguish.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.