Learn More
BUCHI Printer Paper for Melting Point M-562 Apparatus
Product Code. 10260642 Shop All Buchi Products
Click to view available options
For Use With (Equipment):
Melting Point M-562 Apparatus
Unit Size:
Each
Product Code. 10260642
Brand: BUCHI 028261
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
For Melting Point M-562 Apparatus.
For Melting Point M-562 Apparatus.
Specifications
Product Type Printer paper
For Use With (Equipment) Melting Point M-562 Apparatus