Learn More
Witeg™ Buechner Filter Funnels
For use in general filtration applications in the laboratory.
31.40€ - 39.05€
Specifications
Material | Borosilicate Glass |
---|---|
For Use With (Application) | Filtration |
Length (Metric) Stem | 10 mm |
Product Type | Buechner filter funnel |
Product Code | Brand | Capacity (Metric) | Pore Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Pore Size | Price | Quantity & Availability | ||||
11857810
|
Witeg™
2 367 053 |
50 mL | P3 |
31.40€
Each |
Estimated Shipment: 14-05-2024
Log in to see stock. |
||||
11877810
|
Witeg™
2 367 073 |
75 mL | P3 |
35.30€
Each |
Estimated Shipment: 24-05-2024
Log in to see stock. |
||||
11887810
|
Witeg™
2 367 074 |
75 mL | P4 |
39.05€
Each |
Estimated Shipment: 16-05-2024
Log in to see stock. |
||||
Description
- Made from borosilicate glass
- Multiple capacities available
- Multiple disc sizes and stem lengths
Specifications
Borosilicate Glass | |
Filtration | |
10 mm | |
Buechner filter funnel |