missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer solution pH 4.008 (25°C), With fungicide, according to DIN 19266, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
1 L
Unit Size:
1 L
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| CAS | 1336-21-6 |
| pH | 4.008 |
| Quantity | 1 L |
| Molecular Formula | H5NO |
| MDL Number | MFCD00066650 |
| UN Number | NONH for all modes of transport |
| InChI Key | VHUUQVKOLVNVRT-UHFFFAOYSA-N |
| SMILES | N.O |
| IUPAC Name | azanium;hydroxide |
| Molecular Weight (g/mol) | 35.05 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction