missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution, pH 10.105, Eu. Ph., Chem-Lab

Product Code. 11943353
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
This item is not returnable. View return policy

Product Code. 11943353

Brand: Chem-Lab CL07.0208.0500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Reference buffer for laboratory use. Meets specifications of Eur. Ph. 4(2.2.3) for pH measurements. Traceable to SRM 191b and 192b of NIST.

pH = 10.32 at 0°C
pH = 10.25 at 5°C
pH = 10.16 at 10°C
pH = 10.12 at 15°C
pH = 10.06 at 20°C
pH = 10.01 at 25°C
pH = 9.97 at 30°C
pH = 9.93 at 35°C
pH = 9.89 at 40°C
pH = 9.86 at 45°C
pH = 9.83 at 50°

Specifications

Density 1.00g/mL
pH 10.015
Quantity 500 mL
Packaging PE
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.