Learn More
Buffer Solution pH 10, Chem Lab™
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Specifications
Specifications
| Melting Point | 0°C |
| Color | Colorless |
| pH | 10 |
| Physical Form | Liquid |
| Quantity | 500 mL |
| Solution Type | Buffer |
| Formulation | 54g NH4Cl + 233mL NH4OH 25%/H2O |
| Odor | Ammonia |
| Solubility Information | Water Soluble |