missing translation for 'onlineSavingsMsg'
Learn More

Buffer solution pH 4.008 (25°C), With fungicide, according to DIN 19266, Honeywell Fluka™

Product Code. 15684010 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
6 x 1 L
Unit Size:
Pack of 6
This item is not returnable. View return policy

Product Code. 15684010

Brand: Honeywell-Fluka 335936X1L

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

With fungicide, according to DIN 19266

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Buffer solution pH 4.008 (25°C)
Name Note With fungicide, according to DIN 19266
CAS 877-24-7, 7732-18-5, 100-56-1
Boiling Point 100°C
CAS Min % >1.0000%, >95.0000%
CAS Max % <5.0000%, <100.0000%
Density 1.0 g/cm3
Quantity 6 x 1 L
Solution Type Buffer
MDL Number MFCD00081185
Packaging Poly Bottle
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.