missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer solution pH 7.0 (20°C), Green colored, potassium dihydrogen phosphate / disodium hydrogen phosphate, with fungicide, Honeywell Fluka™
Green colored, potassium dihydrogen phosphate / disodium hydrogen phosphate, with fungicide
Brand: Honeywell Fluka 33666-6X500ML
Additional Details : Poids : 2.98500kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Buffer solution pH 7.0 (20°C) | |
≥90.0000% | |
7778-77-0 | |
Potassium sodium phosphate buffer solution | |
Poly Bottle | |
6 x 500mL |
Green colored, potassium dihydrogen phosphate / disodium hydrogen phosphate, with fungicide | |
≤1.0000% | |
MFCD00146206 | |
Buffer Solution | |
1.0 g/cm3 |