missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution, pH 7, Potassium Dihydrogen Phosphate/Sodium Hydroxide, Honeywell™ Fluka™

Product Code. 15666930 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
1 L
Unit Size:
1 L
This item is not returnable. View return policy

Product Code. 15666930

Brand: Honeywell Fluka 732081L

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 1336-21-6
Density 1.0g/mL at 20°C
pH 7
Quantity 1 L
Molecular Formula H5NO
MDL Number MFCD00066650
Packaging Poly bottle
UN Number NONH for all modes of transport
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
SMILES N.O
IUPAC Name amine hydrate
Molecular Weight (g/mol) 35.05
PubChem CID 14923
ChEBI CHEBI:18219
Chemical Name or Material Buffer solution, (potassium dihydrogen phosphate/sodium hydroxide)
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.