Learn More
Buffer Solution, pH 9.00, Colored (Green), NIST, Chem-Lab
Catalog Number 11943083
Click to view available options
Quantity:
5 L
Packaging:
EP
H3BO3-KCl-NaOH buffer solution (green) for laboratory use and pH measurements, traceable to SRM from NIST
pH = 9.24 at 0°C
pH = 9.11 at 10°C
pH = 9.00 at 20°C
pH = 8.97 at 25°C
pH = 8.93 at 30°C
pH = 8.86 at 40°C
pH = 8.80 at 50°C
pH = 8.75 at 60°C
pH = 8.71 at 70°C
pH = 8.67 at 80°C
pH = 8.64 at 90°C
Color Green
Density 1.00g/mL
pH 9.00
Quantity 5 L
Formulation H3BO3, KCl, NaOH
Packaging EP
Traceability to NIST NIST traceable