missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bumetrizole, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | Bumetrizole |
| CAS | 3896-11-5 |
| Molecular Formula | C17 H18 Cl N3 O |
| Quantity | 10 g |
| Synonym | p-Cresol, 2-tert-butyl-6-(5-chloro-2H-benzotriazol-2-yl)- (7CI,8CI), 2-(5-Chloro-2H-benzotriazol-2-yl)-6-(1,1-dimethylethyl)-4-methylphenol, 2-(2-Hydroxy-3-tert-butyl-5-methylphenyl)-5-chloro-2H-benzotriazole, 2-(2-Hydroxy-3-tert-butyl-5-methylphenyl)-5-chlorobenzotriazole, 2-(2'-Hydroxy-3'-tert-butyl-5'-methylphenyl)-5-chlorobenzotriazole, 2-(2'-Hydroxy-3'-tert-butyl-5'-methylphenyl)-5-chlorobenztriazole, 2-(3-tert-Butyl-2-hydroxy-5-methylphenyl)-5-chloro-2H-benzo-v-triazole, 2-(3-tert-Butyl-2-hydroxy-5-methylphenyl)-5-chloro-2H-benzotriazole, 2-(3-tert-Butyl-2-hydroxy-5-methylphenyl)-5-chlorobenzotriazole, 2-(3-tert-Butyl-5-methyl-2-hydroxyphenyl)-5-chlorobenzotriazole, 2-(3'-tert-Butyl-2'-hydroxy-5'-methylphenyl)-5-chlorobenzotriazole, 2-(3'-tert-Butyl-5'-methyl-2'-hydroxyphenyl)-5-chlorobenzotriazole, 2-(5-Chloro-2-benzotriazolyl)-6-tert-butyl-p-cresol, 2-(5-Chloro-2H-benzotriazol-2-yl)-6-tert-butyl-4-methylphenol, 2-[5-Chloro-2H-benzotriazol-2-yl]-4-methyl-6-(tert-butyl)phenol, 2-tert-Butyl-6-(5-chloro-2-benzotriazolyl)-4-methylphenol, 2-tert-Butyl-6-(5-chloro-2H-benzotriazol-2-yl)-4-methylphenol, 5-Chloro-2-(2-hydroxy-3-tert-butyl-5-methylphenyl)-2H-benzotriazole, 5-Chloro-2-(3-tert-butyl-2-hydroxy-5-methylphenyl)benzotriazole, A-Light, ADK Stab LA 36, ADK Stab LA 36RG, Benazol PBKh, Bumetrizole, CBT, Cibatex LF, Eversorb 73, J 395, J 395 (light stabilizer), JC 30S, JF 79, Kemisorb 73, LA 36, Light Stabilizer 326, Lowilite 26, Mark LA 36, Seesorb 703, Songsorb 3260, Sumisorb 300, T 326, TNV 326, Tin 326, Tinogard AS, Tinuvin 326, Tomisorb 600, UV 326, Uvinul 3026, VS 550, Viosorb 550 |
| InChI Formula | InChI=1S/C17H18ClN3O/c1-10-7-12(17(2,3)4)16(22)15(8-10)21-19-13-6-5-11(18)9-14(13)20-21/h5-9,22H,1-4H3 |
| SMILES | Cc1cc(c(O)c(c1)C(C)(C)C)n2nc3ccc(Cl)cc3n2 |
| IUPAC Name | 2-tert-butyl-6-(5-chlorobenzotriazol-2-yl)-4-methylphenol |
| Molecular Weight (g/mol) | 315.797 |
| Formula Weight | 315.1138 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction