Learn More
Aalborg™ Buna N O-ring Aluminum Frame with Plug
Aalborg™ Buna N O-ring Aluminum Frame with Plug is for use with multiple tube flow meters, which retain most of the unique features associated with single tube units.
259.70€ - 500.32€
Specifications
Type Frame
View More Specs
Products 7
Product Code Brand For Use With (Equipment) Includes Price Quantity & Availability  
Product Code Brand For Use With (Equipment) Includes Price Quantity & Availability  
11768528
View accessory products View Documents Promotion Details
Aalborg™
P21A2-BA0
2-Tube 150 mm Parallel Mounting Gas/Liquid Flowmeter Plug
259.70€
Each
Estimated Shipment: 09-01-2025
to see stock.
Add to basket
11778528
View accessory products View Documents Promotion Details
Aalborg™
P21A2-BA2
2-Tube 150 mm Manifold Mounting Gas/Liquid Flowmeter Plug and Manifold
296.80€
Each
Estimated Shipment: 18-12-2024
to see stock.
Add to basket
11748548
View accessory products View Documents Promotion Details
Aalborg™
P41A2-BA0
4-Tube 150 mm Parallel Mounting Gas/Liquid Flowmeter Plug
439.90€
Each
Estimated Shipment: 06-12-2024
to see stock.
Add to basket
11753189
View accessory products View Documents Promotion Details
Aalborg™
P41A2-BA2
4-Tube 150 mm Manifold Mounting Gas/Liquid Flowmeter Plug and Manifold
500.32€
Each
Estimated Shipment: 15-01-2025
to see stock.
Add to basket
11753349
View accessory products View Documents Promotion Details
Aalborg™
P51A2-BA2
5-Tube 150 mm Manifold Mounting Gas/Liquid Flowmeter Plug and Manifold
435.66€
Each
Estimated Shipment: 06-12-2024
to see stock.
Add to basket
11728538
View accessory products View Documents Promotion Details
Aalborg™
P31A2-BA0
3-Tube 150 mm Parallel Mounting Gas/Liquid Flowmeter Plug
336.02€
Each
Estimated Shipment: 15-01-2025
to see stock.
Add to basket
11738538
View accessory products View Documents Promotion Details
Aalborg™
P31A2-BA2
3-Tube 150 mm Manifold Mounting Gas/Liquid Flowmeter Plug and Manifold
386.90€
Each
Estimated Shipment: 15-01-2025
to see stock.
Add to basket
Description
Description
  • Rib-guided or fluted metering tubes facilitate stable, accurate readings
  • Interchangeability of flow tubes and floats
  • Manifolding at inlet or outlet
  • Ease of installation and exchange of flow tubes
  • Chemical compatibility
  • Simple means of mounting
Specifications
Specifications
Frame