Learn More
Abeba™ Business Lady 3980 Shoes
Comfortable with smooth leather upper. Antistatic.
Brand: Abeba™ 3980/42
Additional Details : Weight : 0.70200kg
Description
- Smooth leather
- Black
- Antistatic leather outsole sole
- ESD dissipative according to DIN EN 61340
- Lining microfiber, respiration absorbing
- Instep area with lacing
- Heel 60 mm
Specifications
Black | |
Leather | |
Leather |
Microfiber | |
42 | |
3980 |