Learn More
Abeba™ Business Men 32430 Shoes
ESD dissipative with high wearing comfort. Smooth leather construction.
Brand: Abeba™ 32430/41
Additional Details : Weight : 1.30000kg
Description
- CE EN ISO 20347:2012 O1 SRA
- Smooth leather
- Black
- Slip resistance special rubber outsole sole
- ESD dissipative according to DIN EN 61340
- Lining microfiber, respiration absorbing
- Instep area with lacing
- Insole leather
Specifications
CE EN ISO 20347:2012 O1 SRA | |
Microfiber | |
41 | |
Rubber |
Black | |
Leather | |
Slip-resistant | |
32430 |