Learn More
Abeba™ Business Men 32430 Shoes
ESD dissipative with high wearing comfort. Smooth leather construction.
Brand:  Abeba™ 32430/43
Additional Details : Weight : 1.30000kg
 View more versions of this product
Product Code. 15659677
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
Description
Description
  • CE EN ISO 20347:2012 O1 SRA
  • Smooth leather
  • Black
  • Slip resistance special rubber outsole sole
  • ESD dissipative according to DIN EN 61340
  • Lining microfiber, respiration absorbing
  • Instep area with lacing
  • Insole leather
Specifications
Specifications
CE EN ISO 20347:2012 O1 SRA
Microfiber
43
Rubber
Black
Leather
Slip-resistant
32430