Learn More
Abeba™ Business Men 32450 Shoes
ESD dissipative with high wearing comfort. Smooth leather construction.
Brand:  Abeba™ 32450/47
Additional Details : Weight : 1.36000kg
 View more versions of this product
Product Code. 15659697
  • 92.75€ / 1 pair
Estimated Shipment
22-09-2020
Add to basket
Description
Description
  • CE EN ISO 20347:2012 O1 SRA
  • Smooth leather
  • Black
  • Slip resistance special rubber outsole sole
  • ESD dissipative according to DIN EN 61340
  • Lining microfiber, respiration absorbing
  • Instep area with lacing
  • Insole leather
Specifications
Specifications
CE EN ISO 20347:2012 O1 SRA
Microfiber
47
Rubber
Black
Leather
Slip-resistant
32450