Learn More
Abeba™ Business Men 33240 Shoes
ESD dissipative with high wearing comfort. Smooth leather construction.
Brand:  Abeba™ 33240/36
Additional Details : Weight : 1.12000kg
 View more versions of this product
Product Code. 15659817
  • 161.06€ / 1 pair
Estimated Shipment: 25-04-2024
to see stock.
Add to basket
Description
Description
  • CE EN ISO 20345:2011 S2 SRA
  • Smooth leather
  • Black
  • Steel toe cap
  • Slip resistance special rubber outsole sole
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Lining microfiber, respiration absorbing
  • Instep area with lacing
Specifications
Specifications
CE EN ISO 20345:2011 S2 SRA
Microfiber
36
Rubber
Steel
Black
Leather
Slip-resistant
33240