Learn More
Abeba™ Business Men 33240 Shoes
ESD dissipative with high wearing comfort. Smooth leather construction.
Brand: Abeba™ 33240/46
Additional Details : Weight : 1.12000kg
Description
- CE EN ISO 20345:2011 S2 SRA
- Smooth leather
- Black
- Steel toe cap
- Slip resistance special rubber outsole sole
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Lining microfiber, respiration absorbing
- Instep area with lacing
Specifications
CE EN ISO 20345:2011 S2 SRA | |
Microfiber | |
46 | |
Rubber | |
Steel |
Black | |
Leather | |
Slip-resistant | |
33240 |