Learn More
Butyl 4-hydroxybenzoate-ring-13C6 solution, 50 μg/mL in acetone, Honeywell Fluka™
Product Code. 15990182 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Product Code. 15990182
Brand: Honeywell Fluka R13491ML
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
50 μg/mL in acetone. Analytical standard, reference material
Ensure you have the information you need to quickly start your analysis, and pass relevant audits:
  • High quality inorganic and organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results
  • Free expert technical support
TRUSTED_SUSTAINABILITY
Specifications
Packaging Glass Bottle
CAS 1416711-53-9, 67-64-1
Density 0.79 g/cm3
Flash Point -17°C
Physical Form Liquid
Boiling Point 55°C to 57°C
Industry Type Parabenes
Quantity 1 mL
Grade Analytical Standard