missing translation for 'onlineSavingsMsg'
Learn More

Butyl acrylate, 99+%, stabilized, Thermo Scientific Chemicals

Catalog Number p-3735870
36.20 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
1 L
2.5 L
10 L
Packaging:
Glass Bottle
Plastic drum
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
141-32-2, 150-76-5
C7H12O2
128.17
MFCD00009446
CQEYYJKEWSMYFG-UHFFFAOYSA-N
butyl acrylate, n-butyl acrylate, acrylic acid butyl ester, n-butyl propenoate, 2-propenoic acid, butyl ester, butyl 2-propenoate, butylacrylate, acrylic acid, butyl ester, acrylic acid n-butyl ester, butylester kyseliny akrylove
8846
CHEBI:3245
butyl prop-2-enoate
CCCCOC(=O)C=C
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
141-32-2, 150-76-5
C7H12O2
128.17
MFCD00009446
CQEYYJKEWSMYFG-UHFFFAOYSA-N
butyl acrylate, n-butyl acrylate, acrylic acid butyl ester, n-butyl propenoate, 2-propenoic acid, butyl ester, butyl 2-propenoate, butylacrylate, acrylic acid, butyl ester, acrylic acid n-butyl ester, butylester kyseliny akrylove
8846
CHEBI:3245
butyl prop-2-enoate
CCCCOC(=O)C=C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Acidity 0.01% max. (as acrylic acid)
Melting Point -64.0°C
Density 0.8900g/mL
Boiling Point 145.0°C
Flash Point 39°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass Bottle
Linear Formula H2C=CHCO2(CH2)3CH3
Refractive Index 1.4170 to 1.4190
Quantity 1 L
Beilstein 02, II, 388
Merck Index 15, 1541
Specific Gravity 0.89
Solubility Information Solubility in water: 1.4g/L (20°C)
Stabilizer 10 to 20ppm MEHQ
Viscosity 0.869 mPa.s (20°C)
Formula Weight 128.17
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Butyl acrylate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'