Learn More
C1orf96 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA560366
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: EEQDAEAGDA EAEDAEDAAL PALPVKDVED KPEQQTRTRE TDKSPTSTEP RQQPSALFAR GNRKAVKSPQ RSSSKIKENK HPFALYGW Highest antigen sequence identity to the following orthologs: Mouse - 63%, Rat - 63%.
C1orf96 Polyclonal antibody specifically detects C1orf96 in Human samples. It is validated for Immunohistochemistry.Specifications
C1orf96 | |
Polyclonal | |
Unconjugated | |
CCSAP | |
1700054N08Rik; AI853424; C1orf96; C4H1orf96; Ccsap; centriole and spindle-associated protein; centriole, cilia and spindle associated protein; centriole, cilia and spindle-associated protein; CSAP; RGD1563235 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
126731 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6IQ19 | |
CCSAP | |
Recombinant protein corresponding to Human C1orf96 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only