Learn More
Consort C3010/C3030 Multiparameter Dual Channel Analyzers
Product Code. 11955448
Click to view available options
Product Type:
Multiparameter Analyzer
Total Meter Kit
pH Meter Kit
Unit Size:
Each
Product Code. 11955448
Brand: Consort C3030T
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Two independent channels for all measurements. Consort C3010/C3030 Multiparameter Dual Channel Analyzers are full parameter benchtop meters with unique measurement hardware for each channel, separate temperature inputs, data logging and a USB/RS232 interface for connection to/control by a PC.
  • Multichannel up to 2 measurements can be performed at the same time (2X/sec)
  • Multiparameter for pH, mV, conductivity, resistivity, salinity, TDS, dissolved oxygen, temperature, ion (3030 only), and air pressure
  • No interference between pH/ORP/Ion and conductivity electrodes in the same solution
  • Custom calibration tables for buffer/standard-temperature relations (via device menu or from PC)
  • Wide display shows 2 channels at the same time, including temperature and date/time
  • Stability indicator ensures visualization when the measurement has stabilized
  • Stability algorithm ensures stable readings with ability to detect fast changes
  • Gold plated BNC connectors each accept different electrodes; gold plating prevents corrosion
  • Hold function allows to freeze the display
  • Selectable resolution for more stable readings of mV, pH and DO
  • Range lock for conductivity measurements
  • Built-in help system helps guide users through all steps of calibration and use
  • Capacitive compensation eliminates electrode/cable capacitive component at low conductivities
  • Galvanic isolated USB interface eliminates ground loop effects when connected to a PC
  • GPL report can be shown on the display or sent to the digital port
  • Pre-programmed standards:
    pH: 1.68, 2.00, 4.00, 4.01, 6.87, 6.99, 9.18, 9.21, 10.01, 12.00, 12.45 (at 25°C)
    Conductivity: 1413 μS/cm, 12.88 mS/cm, 111.8 mS/cm (at 25°C)
  • Free software and firmware updates downloadable from www.consort.be
Specifications
Outputs USB/RS232
Accuracy (Conductivity) 0.5% f.s. of range
Accuracy (DO) 1% ± 1 digit
Accuracy (ISE) 0.5% ± 1 digit
Accuracy (mV) 0.1% ± 1 digit
Accuracy (pH) 0.1% ±1 digit
Backlight Option White back light
Calibration GLP
Calibration Points 1 (mV, DO, temperature, air pressure)
Calibration Points (Conductivity) 1 to 3
Calibration Points (ISE) 2 to 5 points + blank
Calibration Points (pH) 1 to 5
Cell Constant 0.07 to 13 cm-1
Data Management Details Measurement + ° C/date/time
Data Points 12,000
Channels 2 measurement, 2 temperature
Display Type LCD, 240 x 64 pixel
Range (Conductivity) 0 to 2000 mS/cm
Range (mV) ±2000.0 mV
Range (pH) -2.000 to 16.000
Range (Resistivity) 0 to 200 Mohm/cm
Range (Salinity) 0.0 to 70.0
Range (TDS) 0.0 to 100.0 g/L
Resolution (ISE) 3 digits
Resolution (pH) 0.001
Temperature Range (Metric) -5.0°C to +105.0°C
For Use With (Application) ion, pH, mV, conductivity, resistivity, salinity, TDS, DO, air pressure, temperature
Includes Compete Meter Kit: C3030 Meter, pH/ATC electrode, conductivity/ATC electrode, buffers (pH 4 and 7), electrolyte (3 m KCl), conductivity standard (0.01 m KCl), DO electrode
Inputs 2 BNC (measurement), 2 x 2 banana (for Pt1000 for temperature)
Languages English, Dutch, French, German
Model C3030
Pressure Range 600 - 1300 hPa
Product Type Total Meter Kit
Range (DO) 0.00 - 60.00 mg/L
Range (ISE) 0.01 ng/L to 100 g/L
Relative Humidity Range 0 - 95%, non-condensing
Resolution (Conductivity) 0.001 μS/cm
Resolution (DO) 0.01 mg/L
Resolution (mV) 0.1 mV
Resolution (Resistivity) 1 Ω.cm
Resolution (TDS) 0.01 mg/L
Salinity Factor Ref temp: 15° C
Temperature Accuracy 0.1°C
Temperature Compensation −5.0° to +105.0° C (pH, conductivity), 0° to 50° C (DO)
Temperature (Metric) Operating 0°C to 40°C
Temperature (Metric) Reference 20°/25° C (conductivity)
Temperature Resolution 0.1° C
Warranty 36 Months
Dimensions (L x W x H) 26W x 18D x 9H cm
Weight (Metric) 1 kg
Voltage 100/240 V
Frequency 50/60 Hz