Learn More
YSI™ Cables and DO/Temp/Conductivity Probes for 556 Multi-Probe System
Click to view available options
Length (Metric) Cable:
10 m
20 m
4 m
Length (English) Cable:
13 ft.
32 ft.
64 ft.
Unit Size:
Each
Description
DO/Temperature/Conductivity Probe and Cable. Required to measure dissolved oxygen, temperature, conductivity.
Specifications
Specifications
| Length (English) Cable | 13 ft. |
| Length (Metric) Cable | 4 m |
| Includes | Temperature/conductivity sensor |
| Warranty | 1 Year |
| For Use With (Equipment) | YSI model 556 handheld multi probe meter |
| For Use With (Application) | Measures DO, temperature, conductivity |