Learn More
Cadence Science™ Micro-Emulsifying Needles
Double-hub needles are designed for use in mixing two liquid media differing in viscosity—especially for adjuvants and antigens, oil-based liquids, allergens
Brand: Cadence Science™ 7978
Additional Details : Weight : 0.80000kg
Description
Double-hub needles are designed for use in mixing two liquid media differing in viscosity—especially for adjuvants and antigens, oil-based liquids, allergens
- Available with or without a reinforcing bar
- Overall length: 2 7/8 in. (7.3cm), except for 14-825-17K and -17L needles which have a length of 1 7/8 in. (4.8cm)
Specifications
Glass | |
For allergens and the mixing of adjuvants and antigens, oil based liquids or any two liquid media differing in viscosity, with reinforcing bar | |
1.87 in. | |
47.5 cm |
20 G | |
Without reinforcing bar | |
Micro Emulsifying Needle w/o Reinforcing Bar | |
Non-sterile |