Learn More
Epredia™ Lab Vision™ Pan Cadherin Rabbit Polyclonal Antibody with BSA and azide
Ensure accurate, reproducible results in immunohistochemistry and western blotting experiments with Epredia™ Cadherin, Pan, Rabbit Polyclonal Antibody.
149.94€ - 698.70€
Specifications
Antigen | Cadherin, Pan |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
12603387
|
Epredia™
RB-9036-P0 |
Concentrated | 100 μL |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
|||||
12683377
|
Epredia™
RB-9036-P1 |
Concentrated | 500 μL |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
|||||
12613387
|
Epredia™
RB-9036-P |
Concentrated | 1 mL |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
|||||
12643387
|
Epredia™
RB-9036-R7 |
Prediluted | 7 mL |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
|||||
Description
Cadherins are calcium-dependent cell adhesion molecules, which play important role in the growth and development of cells via the mechanisms of control of tissue architecture and the maintenance of tissue integrity. In adhesion junctions, cadherins are bound to Beta and gamma catenins, which in turn bind to alpha catenin, an actin binding protein. Cadherins play important role in tumor invasion and metastasis. This antibody is excellent for staining of formalin-fixed, paraffin-embedded tissues.Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human, Mouse, Rat, Chicken, Dog, Monkey, Cow and Xenopus.
Epitope: C-terminal
Immunogen: A synthetic peptide from the C-terminus of chicken N-cadherin
Molecular Weight: 120kDa
Positive Control: MCF-Cells, Tonsil and squamous epithelium
Cellular Localization: Membrane
This antibody is excellent for staining of formalin-fixed, paraffin-embedded tissues.
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
Note:
USA: RUO; Int'l: RUO
Specifications
Cadherin, Pan | |
Polyclonal | |
Rabbit | |
Junction Protein | |
A synthetic peptide from the C-terminus of chicken N-cadherin | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
Bovine, Canine, Chicken, Human, Mouse, Non-human Primate, Rat | |
IgG |