missing translation for 'onlineSavingsMsg'
Learn More

Cafestol, Thermo Scientific Chemicals

Catalog Number 15406859
335.35 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
50 mg
100 mg
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
469-83-0
C20H28O3
316.44
MFCD01075769
DNJVYWXIDISQRD-HWUKTEKMSA-N
cafestol, cafesterol, ccris 1518, unii-ac465t6q6w, 5a,8-methano-5ah-cyclohepta 5,6 naphtho 2,1-b furan-7-methanol, 3b,4,5,6,7,8,9,10,10a,10b,11,12-dodecahydro-7-hydroxy-10b-methyl-, 3bs,5as,7r,8r,10ar,10bs, 5a,8-methano-5ah-cyclohepta 5,6 naphtho 2,1-b furan-7-methanol, 3b,4,5,6,7,8,9,10,10a,10b,11,12-dodecahydro-7-hydroxy-10b-methyl-, 3bs-3balpha,5abeta,7beta,8beta,10aalpha,10bbeta, --cafestol, 1s,4s,12s,13r,16r,17r-17-hydroxymethyl-12-methyl-8-oxapentacyclo 14.2.1.0 1,13 .0 4,12 .0 5,9 nonadeca-5 9 ,6-dien-17-ol, 3bs,5as,7r,8r,10ar,10bs-7-hydroxymethyl-10b-methyl-3b,4,5,6,7,8,9,10,10a,10b,11,12-dodecahydro-5a,8-methanocyclohepta 5,6 naphtho 2,1-b furan-7-ol
108052
CHEBI:3291
(1S,4S,12S,13R,16R,17R)-17-(hydroxymethyl)-12-methyl-8-oxapentacyclo[14.2.1.0¹,¹³.0⁴,¹².0⁵,⁹]nonadeca-5(9),6-dien-17-ol
C[C@@]12CCC3=C(C=CO3)[C@H]1CC[C@@]13C[C@@H](CC[C@@H]21)[C@@](O)(CO)C3
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
469-83-0
C20H28O3
316.44
MFCD01075769
DNJVYWXIDISQRD-HWUKTEKMSA-N
cafestol, cafesterol, ccris 1518, unii-ac465t6q6w, 5a,8-methano-5ah-cyclohepta 5,6 naphtho 2,1-b furan-7-methanol, 3b,4,5,6,7,8,9,10,10a,10b,11,12-dodecahydro-7-hydroxy-10b-methyl-, 3bs,5as,7r,8r,10ar,10bs, 5a,8-methano-5ah-cyclohepta 5,6 naphtho 2,1-b furan-7-methanol, 3b,4,5,6,7,8,9,10,10a,10b,11,12-dodecahydro-7-hydroxy-10b-methyl-, 3bs-3balpha,5abeta,7beta,8beta,10aalpha,10bbeta, --cafestol, 1s,4s,12s,13r,16r,17r-17-hydroxymethyl-12-methyl-8-oxapentacyclo 14.2.1.0 1,13 .0 4,12 .0 5,9 nonadeca-5 9 ,6-dien-17-ol, 3bs,5as,7r,8r,10ar,10bs-7-hydroxymethyl-10b-methyl-3b,4,5,6,7,8,9,10,10a,10b,11,12-dodecahydro-5a,8-methanocyclohepta 5,6 naphtho 2,1-b furan-7-ol
108052
CHEBI:3291
(1S,4S,12S,13R,16R,17R)-17-(hydroxymethyl)-12-methyl-8-oxapentacyclo[14.2.1.0¹,¹³.0⁴,¹².0⁵,⁹]nonadeca-5(9),6-dien-17-ol
C[C@@]12CCC3=C(C=CO3)[C@H]1CC[C@@]13C[C@@H](CC[C@@H]21)[C@@](O)(CO)C3

Natural extract from coffee beans which induces glutathione S-transferase

Natural extract from coffee beans which induces glutathione S-transferase

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 156°C to 158°C
Color White
Density 1.23
Quantity 100 mg
Merck Index 14,1634
Solubility Information Soluble in dichlormethane, DMSO or ethanol
Formula Weight 316.44
Physical Form Powder
Chemical Name or Material Cafestol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'