Learn More
Merck Millipore Calbiochem™ Protease Inhibitor Cocktail Set III, Animal-Free
Protease Inhibitor Cocktail Set III, Animal-Free, is a cocktail of six protease inhibitors with broad specificity for the inhibition of aspartic, cysteine & serine proteases & aminopeptidases.
Brand:  Merck Millipore 535140-1SET
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 14865751
  • 308.00€ / 1 set
Estimated Shipment date
from Supplier 07-07-2020
Add to basket
Description
Description
A cocktail of six protease inhibitors with broad specificity for the inhibition of aspartic, cysteine, and serine proteases as well as aminopeptidases. This cocktail is recommended for use with mammalian cell and tissue extracts. Each vial contains 100mm AEBSF, HCl (Cat. No. 101500), 80μM Aprotinin (Cat. No. 616371), 5mm Bestatin (Cat. No. 200484), 1.5mm E-64 (Cat. No. 324890), 2mm Leupeptin Hemisulfate (Cat. No. 108975) and 1mm Pepstatin A (Cat. No. 516482). Available as 1mL vial or a set of 5 x 1 ml.
Specifications
Specifications
In 1mL DMSO.
Avoid Freeze-Thaw Cycles
Aspartic, cysteine, and serine proteases as well as aminopeptidases
Liquid
1 set