Learn More
Merck Millipore Calbiochem™ Protease Inhibitor Cocktail Set III, EDTA-Free
Protease inhibitor cocktail III, EDTA-free for inhibiting aspartic, cysteine, and serine proteases as well as aminopeptidases in mammalian cells and tissues.
Brand:  Merck Millipore 539134-1ML
Additional Details : Weight : 0.00100kg
 View more versions of this product
Product Code. 12818395
  • 94.38€ / 1mL
Estimated Shipment
16-06-2021
Add to basket
Description
Description
A cocktail of six protease inhibitors with broad specificity for the inhibition of aspartic, cysteine, and serine proteases as well as aminopeptidases. This cocktail is recommended for use with mammalian cell and tissue extracts, but is also suitable for bacterial extracts. Supplied with a data sheet.
Specifications
Specifications
Liquid
1mL
Aspartic, cysteine, and serine proteases as well as aminopeptidases