missing translation for 'onlineSavingsMsg'
Learn More

Calcein, TRC

Product Code. 30347509
Click to view available options
Quantity:
10 g
Unit Size:
10g
This item is not returnable. View return policy

Product Code. 30347509

Brand: Toronto Research Chemicals TRCC12541310G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Calcein
CAS 1461-15-0
Molecular Formula C30 H26 N2 O13
Quantity 10 g
Synonym Fluorexon, Oftasceine, Fluorescein complexon, 2,7-Bis[N,N-bis(carboxymethyl)aminomethylene]fluorescein
InChI Formula InChI=1S/C30H26N2O13/c33-21-7-23-19(5-15(21)9-31(11-25(35)36)12-26(37)38)30(18-4-2-1-3-17(18)29(43)45-30)20-6-16(22(34)8-24(20)44-23)10-32(13-27(39)40)14-28(41)42/h1-8,33-34H,9-14H2,(H,35,36)(H,37,38)(H,39,40)(H,41,42)
SMILES OC(=O)CN(CC(=O)O)Cc1cc2c(Oc3cc(O)c(CN(CC(=O)O)CC(=O)O)cc3C24OC(=O)c5ccccc45)cc1O
IUPAC Name 2-[[7'-[[bis(carboxymethyl)amino]methyl]-3',6'-dihydroxy-3-oxospiro[2-benzofuran-1,9'-xanthene]-2'-yl]methyl-(carboxymethyl)amino]acetic acid
Molecular Weight (g/mol) 622.53
Formula Weight 622.1435
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.