missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium chloride-2-hydrate, <0.0001% AL, Honeywell™
Click to view available options
Quantity:
25 kg
Packaging:
Paper / Poly Bag
Specifications
CAS | 10035-04-8 |
CAS Max % | 100.0000% |
Quantity | 25 kg |
Packaging | Paper / Poly Bag |
Chemical Name or Material | Calcium chloride-2-hydrate, <0.0001% AL |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'