Learn More
Calcium iodide, ultra dry, 99.995% (metals basis), Thermo Scientific™
177.00€
Chemical Identifiers
| CAS | 10102-68-8 |
|---|---|
| Molecular Formula | CaI2 |
| Molecular Weight (g/mol) | 293.887 |
| MDL Number | MFCD00010910 |
| InChI Key | UNMYWSMUMWPJLR-UHFFFAOYSA-L |
| Synonym | calcium iodide, calcium diiodide, calciumiodide, calcium 2+ diiodide, acmc-1c61l, ksc491a2n, calcium iodide, anhydrous 25g |
| PubChem CID | 5462722 |
| ChEBI | CHEBI:81718 |
| IUPAC Name | calcium;diiodide |
| SMILES | [Ca+2].[I-].[I-] |
Beskrivning
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Ampouled under argon
Kemiska identifierare
| 10102-68-8 | |
| 293.887 | |
| UNMYWSMUMWPJLR-UHFFFAOYSA-L | |
| 5462722 | |
| calcium;diiodide |
| CaI2 | |
| MFCD00010910 | |
| calcium iodide, calcium diiodide, calciumiodide, calcium 2+ diiodide, acmc-1c61l, ksc491a2n, calcium iodide, anhydrous 25g | |
| CHEBI:81718 | |
| [Ca+2].[I-].[I-] |
Specifikationer
| 10102-68-8 | |
| MFCD00010910 | |
| calcium iodide, calcium diiodide, calciumiodide, calcium 2+ diiodide, acmc-1c61l, ksc491a2n, calcium iodide, anhydrous 25g | |
| UNMYWSMUMWPJLR-UHFFFAOYSA-L | |
| calcium;diiodide | |
| 5462722 | |
| 293.89 | |
| Odorless | |
| Hygroscopic; Light sensitive | |
| (metals basis) |
| CaI2 | |
| 14,1677 | |
| Soluble in water (64.6g/100 mL (0°C)). Soluble in acetone and alcohols. | |
| [Ca+2].[I-].[I-] | |
| 293.887 | |
| CHEBI:81718 | |
| 99.995% | |
| Ampule | |
| 3.956 g/mL | |
| Calcium iodide, Ultra dry |
Säkerhet och hantering
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
missing translation for 'einecsNumber' : 233-276-8
missing translation for 'tsca' : Yes
missing translation for 'storageNote' :
- Ampouled under argon
Rekommenderad förvaring : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.