missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Sulfate Dihydrate, Certified AR for Analysis, Fisher Chemical™
Shop All Fisher Chemical Products
Click to view available options
Quantity:
250 g
500 g
1 kg
Unit Size:
1kg
250g
500g
Specifications
Specifications
| Color | White |
| pH | 7 |
| Physical Form | Powder/Solid |
| Quantity | 250 g |
| Formula Weight | 172.17g/mol |
| Packaging | Plastic powder jar |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction