Learn More
BINDER™ Calibration Certificate, Temperature and Humidity
Shop All BINDER Products
Click to view available options
For Use With (Equipment):
BINDER Chambers Series MKF
BINDER Chambers Series MKFT
Calibration Points:
1 Measuring Point
27 Measuring Points
Unit Size:
Each
Beskrivelse
- One humidity point of measurement
- Including certificate
- Humidity measurement point in center of chamber
- Temperature (according to DIN12880) and humidity measurement at 25°C / 60% RH or at specified values
Tekniske data
Tekniske data
| For Use With (Equipment) | BINDER Chambers Series MKFT |
| Description | Calibration certificate, temperature and humidity - Measurement in center of chamber at 25°C / 60% RH or at specified test values |
| Calibration Points | 1 Measuring Point |
| Product Type | Calibration Certificate (Temperature and Humidity) |
| For Use With (Application) | Temperature and Humidity Measurement |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion