Learn More
Agilent BioTek Calibration Solution with 1 Cuvette
Product Code. 16523042 Shop All Agilent BioTek Products
Click to view available options
For Use With (Equipment):
Gen5v1.0 and Take3 Module Only, Agilent BioTek Microplate Washer and Reader Instruments
Unit Size:
Each
Product Code. 16523042
Brand: Agilent BioTek 4690008
Calibration Solution with 1 Cuvette includes the Take3 Calibration Solution and one quartz, stoppered cuvette.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Gen5v1.0 and Take3 Module Only, Agilent BioTek Microplate Washer and Reader Instruments
Includes Take3 Calibration Solution (PN: 4693002); 1 quartz cuvette with stopper (PN: 48723)