Learn More
Thermo Scientific™ Calibration Validation Carousels
Thermo Scientific™ Performance Verification Standards, Automated (CVC) will ensure the accuracy and reliability of your data while saving time and money.
Brand: Thermo Scientific™ 10010721
Additional Details : Weight : 0.10000kg
Description
Thermo Scientific™ Performance Verification Standards, Automated (CVC) will ensure the accuracy and reliability of your data while saving time and money. The CVC provides certified reference materials in a fully automated confirguration for hands-free performance verification of your Thermo Scientific™ spectrophotometer.
- Simplify qualification procedures by automating performance verification of UV-Visible spectrophotometers
- Each CVC has a unique serial number matched automatically to the corresponding data files containing standard values and instrument specifications
- Generated result reports are suitable for printing or archival purposes
- Included standards are traceable to either NIST™ or NPL primary standards
- CVCs are available in standard or pharmacopeia compliant configurations
Comprehensive Performance Verification Testing
- Wavelength Accuracy and Repeatability
- Photometric Accuracy and Repeatability
- Stray Light
- Resolution
- Photometric Noise
- Photometric Stability or Drift
- Baseline Flatness
Save Time, Reduce Errors
- Eliminates need to copy standard values from certificates
- Eliminates need to analyze standards individually
- Eliminates need to match standards to data
- Eliminates need for manual calculations
- Eliminates need for manual transcription and reporting of results
Ordering Information:
Unit is shipped in a plastic case for optimal protection during storage. Calibration Validation Carousels can be used with INSIGHT, INSIGHT Security, VISIONpro™, VISIONsecurity™ and applicable Local Control software. Pharmacopeia-compliant CVCs are compatible with the Evolution 600, 300, 260 Bio, 220 and 201 UV-Visible spectrophotometers. Standard configuration CVCs are additionally compatible with the AquaMate Plus, BioMate 6, Evolution 160, Helios Omega, Helios Zeta and UV-10 UV-Visible spectrophotometers.
Specifications
All standards contained in the CVC are certified in the Thermo Fisher Scientific ISO/IEC 17025 accredited laboratory traceable to the NIST in the United States or the NPL in the United Kingdom. | |
Routine Performance Validation - NPL Traceable Filters | |
Calibration Validation Carousel with Smart Base |
NIST traceable | |
Evolution 201, 220, 260 Bio and 300 | |
10010721 |