missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Calponin-1 Rabbit Monoclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry procedures with Epredia™ Calponin-1, Rabbit Monoclonal Antibody.

190.74€ - 1175.00€

Specifications

Antigen Calponin
Clone EP798Y
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 4
Product Code Brand Form Quantity Price Quantity & Availability  
Product Code Brand Form Quantity Price Quantity & Availability  
12643607
View Documents Promotion Details
Epredia™
RM-2102-S
Concentrated 1 mL
1175.00€
1mL
View alternative products
Save Up to 
»
Estimated Shipment: 17-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
12653607
View Documents Promotion Details
Epredia™
RM-2102-S0
Concentrated 100 μL This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
12623607
View Documents Promotion Details
Epredia™
RM-2102-R7
Prediluted 7 mL This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
12633607
View Documents Promotion Details
Epredia™
RM-2102-RQ
Ready-to-Use 12 mL This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
Description

Description

Calponin, a thin filament associated protein, is implicated in the regulation and modulation of smooth muscle contraction. It is capable of binding to actin, calmodulin, troponin C and tropomyosin. Calponin is expressed in smooth muscle and tissues containing significant amounts of smooth muscle. Two isoforms of calponin exist whose molecular weights are 34 kDa and 29 kDa. Expression of the 29 kDa form is primarily restricted to muscle of the urogenital tract. The expression of calponin has also been demonstrated in myoepithelial cells from benign and malignant breast lesions. It stains smooth muscle, myoepithelial cells, myofibroblasts, keratinocytes and nerve fibers. It identifies myoepithelial cells in breast lesions, and helps differentiate breast collagenous spherulosis (positive) from adenoid cystic carcinoma. Adenoid cystic carcinoma in salivary gland tumors is calponin positive.

Host Species: Rabbit

Clone: EP798Y

Species Reactivity: Human. Others not known.

Immunogen: A synthetic peptide derived from near the C-terminus of human Calponin-1

Molecular Weight: 29/34 kDa

Positive Control: Breast CA

Cellular Localization: Cytoplasm Myoepithelial cells

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Specifications

Calponin
Immunohistochemistry (Paraffin)
Unconjugated
IVD
A synthetic peptide derived from near the C-terminus of human Calponin-1
EP798Y
Monoclonal
Rabbit
Cancer and Tumor Biology
Primary
Videos
SDS
Documents

Documents

Product Certifications