Learn More
Olympus™ Camera Mount Lens, 0.4X
Connect a camera to your microscope and record visualizations. Olympus™ Camera Mount Lens, 0.4X connects to BX, CX, MX, STM, SZX, IX and GX series microscopes.
Brand: Olympus™ K10017119
Additional Details : Weight : 0.80000kg
Description
- Camera adapter for Olympus™ microscopes.
Specifications
Camera Mount Lens | |
Plan achromatic | |
0.4X | |
0.008 to 0.027 |