Learn More
Invitrogen™ Human CAMK1G (CaMKI gamma), GST Tag Recombinant Protein
Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. This CAMK1G (CaMKI gamma) recombinant human protein (full length) was expressed in insect cells. CAMK1G (CaMKI gamma) recombinant human protein is also available in our SelectScreen™ Kinase Profiling Service. Validated for use with the Z'-LYTE™ assay Available in multiple pack sizes for convenient use Also available in our SelectScreen Kinase Profiling Service Protein Form: Full Length, Recombinant, Active The gene accession number for CAMK1G (CaMKI gamma) is NP_065172.1.
Specifications
Specifications
| Accession Number | Q96NX5 |
| Concentration | See Label |
| For Use With (Application) | Kinase Assay |
| Formulation | 50mM tris HCl with 10mM glutathione, 0.1mM EDTA, 25% glycerol, 150mM NaCl, 0.1mM PMSF, 0.25mM DTT and no preservative; pH 7.5 |
| Gene ID (Entrez) | 57172 |
| Molecular Weight (g/mol) | 79.8 kDa |
| Name | Human CAMK1G (CaMKI gamma), GST Tag |
| pH Range | 7.5 |
| Purification Method | Purified |
| Quantity | 1 mg |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.