missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ CAMK2G 293T Cell Transient Overexpression Lysate (Denatured) (T02)

Human overexpression lysate

Brand:  Abnova™ H00000818-T02.100uL

Additional Details : Weight : 0.00010kg


Product Code. 16162571

  • 209.00€ / 100µL
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Western Blot
CAMK2G 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 58.08kDa
Transient overexpression cell lysate was tested with anti-CAMK2G antibody (Mfr. No. H00000818-B02) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
293T
BC034044.1
pCMV-CAMK2G full-length
100 μL
Human
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 100UL CAMK2G 293T Cell Transient Overexpression Ly

Signal Word
  • Warning

Hazard Category
  • Skin sensitiser Category 1

Hazard Statement
  • H317-May cause an allergic skin reaction.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

Supplemental information
  • MIXTURE LIST-Contains : Mercaptoethanol

SDS
Documents

Documents

Certificates
Promotions

Promotions